Learn More
Aggrecan G1-IGD-G2 Domains Mouse anti-Human, Clone: 179509, R&D Systems™
Mouse Monoclonal Antibody
Marke:  RD Systems (Bio-Techne) MAB1220
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13418465
  • 377.53 € / 500 Mikrogramm
Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Aggrecan G1-IGD-G2 Domains Monoclonal antibody MAB1220 Specifically detects Aggrecan G1-IGD-G2 Domains in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blotting
Spezifikation
Spezifikation
Aggrecan
179509
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
ACAN
Mouse myeloma cell line NS0-derived recombinant human Aggrecan, Val20-Gly675, Accession # NP_037359
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
176
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunohistochemistry, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL
NP_037359
ACAN, AGC1, AGC1SEDK, aggrecan, aggrecan core protein, Cartilage-specific proteoglycan core protein, Chondroitin sulfate proteoglycan 1, Chondroitin sulfate proteoglycan core protein 1, CSPCP, CSPG1, CSPG1aggrecan 1, CSPGCP, large aggregating proteoglycan, MSK16, MSK16AGCAN, SEDK
Mouse
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects human Aggrecan G1-IGD-G2 Domains in Western blots and direct ELISAs.