missing translation for 'onlineSavingsMsg'
Learn More

AgRP/ART Mouse anti-Human, Clone: 82815, R&D Systems™

Mouse Monoclonal Antibody

Marke:  RD Systems (Bio-Techne) MAB8312

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13411916

  • 377.53 € / 500 Mikrogramm


Voraussichtlicher Liefertermin 30-04-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

AgRP/ART Monoclonal antibody MAB8312 Specifically detects AgRP/ART in Human samples. It is validated for ELISA Capture
Spezifikation

Spezifikation

AgRP/ART
82815
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
O00253
agouti related protein homolog (mouse), agouti-related protein, Agrt, AGRTMGC118963, ART, ARTagouti (mouse) related protein, ASIP2
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human AgRP in ELISAs and Western blots. In Western blots, this antibody shows 100% cross-reactivity with recombinant mouse AgRP.
Human
ELISA
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
AGRP
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human AgRP Ala21-Thr132 Accession # O00253
Protein A or G purified from ascites
RUO
Primary
181
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos

For Research Use Only