missing translation for 'onlineSavingsMsg'
Learn More

alpha Actinin 4 Monoclonal Antibody (7H6), Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MA1800

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11585372

  • 497.00 € / 100 Mikrogramm
Verfügbar ab: 16-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

MA1-800 detects the alpha Actinin 4 in human and mouse cells. MA1-800 has been successfully used in Western blot, and immunofluorescence procedures. MA1-800 detects alpha Actinin 4 which has a predicted molecular weight of approximately 105 kDa. Under reducing conditions, MA1-800 detects a monomer of alpha Actinin 4 at approximately 52.5 kDa. The MA1-800 immunogen is a synthetic peptide corresponding to residues A(884) P Y Q G P D A V P G A L D(897) of human alpha Actin 4.

alpha Actinin 4 Monoclonal antibody specifically detects alpha Actinin 4 in Human, Mouse samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.
Spezifikation

Spezifikation

alpha Actinin 4
Monoclonal
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
O43707, P57780
Actn4
Synthetic peptide corresponding to residues A(884) P Y Q G P D A V P G A L D(897) of human alpha Actinin 4.
100 μg
Primary
Human, Mouse
Antibody
IgG
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
7H6
Unconjugated
Actn4
actinin alpha 4; ACTININ-4; actinin-Alpha-4; Actn4; alpha actinin 4; alpha-actinin-4; C77391; F-actin cross-linking protein; focal segmental glomerulosclerosis 1; FSGS; FSGS1; non-muscle alpha-actinin 4
Mouse
Protein G
RUO
60595, 81
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote