Learn More
alpha Synuclein Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Marke: Invitrogen PA118258
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Reconstitute in 50 μL of sterile water. Centrifuge to remove any insoluble material. After reconstitution keep aliquots at -20 °C for a higher stability, and at 4 °C with an appropriate antibacterial agent. Glycerol (1:1) may be added for an additional stability. Avoid repetitive freeze/thaw cycles.
alpha Synuclein Polyclonal antibody specifically detects alpha Synuclein in Human, Rat samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
alpha Synuclein | |
Polyclonal | |
Unconjugated | |
SNCA | |
Alpha synuclein; alphaSYN; alpha-synuclein; NACP; non A-beta component of AD amyloid; non-A beta component of AD amyloid; non-A4 component of amyloid; Non-A4 component of amyloid precursor; PARK1; PARK4; PD1; Snca; Syn; synuclein alpha; synuclein alpha-140; synuclein, alpha; synuclein, alpha (non A4 component of amyloid precursor) | |
Sheep | |
Affinity chromatography | |
RUO | |
29219, 6622 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with no preservative | |
P37377, P37840 | |
SNCA | |
Synthetic peptide corresponding to residues P(108) Q E G I L E D M P V D P C(120) of human alpha synuclein protein. | |
50 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |