Learn More
Angiopoietin-like Protein 3/ANGPTL3 Rat anti-Mouse, Clone: 128611, R&D Systems™
Rat Monoclonal Antibody
Marke:  RD Systems (Bio-Techne) MAB13621
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13418585
  • 377.53 € / 500 Mikrogramm
Voraussichtlicher Liefertermin 02-05-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Angiopoietin-like Protein 3/ANGPTL3 Monoclonal antibody MAB13621 Specifically detects Angiopoietin-like Protein 3/ANGPTL3 in Mouse samples. It is validated for ELISA Capture
Spezifikation
Spezifikation
Angiopoietin-like Protein 3/ANGPTL3
128611
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
ANGPTL3
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Angiopoietin-like 3 Ser17-Thr455 Accession # Q9R182
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
27329
Sterile PBS to a final concentration of 0.5 mg/mL.
ELISA
LYOPH
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Q9R182
AGNPT5, Ang-5, angiopoietin 5, Angiopoietin-5, angiopoietin-like 3, Angiopoietin-like protein 3, angiopoietin-related protein 3, ANGPT5angiopoietin-5, ANGPTL3, FHBL2
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Angiopoietin-like 3 in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human ANGPTL3, recombinant mouse (rm) ANGPTL4, or rmAngiopoietin-3 is observed.