Learn More
Angiostatin Kringle 1-3 Mouse anti-Human, Clone: 79735, R&D Systems™
Mouse Monoclonal Antibody
Marke:  RD Systems (Bio-Techne) MAB926
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13412006
  • 420.42 € / 500 Mikrogramm
Voraussichtlicher Liefertermin 29-10-2019
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Description
Description
Angiostatin Kringle 1-3 Monoclonal antibody MAB926 Specifically detects Angiostatin Kringle 1-3 in Human samples. It is validated for Western Blotting
Specifications
Specifications
Angiostatin
79735
Western Blot 1 ug/mL
P00747
PLG
E. coli-derived recombinant human Angiostatin Kringle 1-3 Val98-Ser358 (Cys88Ser, Cys316Ser) Accession # P00747
Protein A or G purified from ascites
RUO
Primary
5340
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Mouse
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Angiostatin Kringle 1-3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody also detects plasminogen but does not cross-react with recombinant mouse Angiostatin.