missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rho GTPase activating protein 1, Mouse, Polyclonal Antibody, Abnova™
Description
Sequence: GFLNIDESQRVPATLQVLQTLPEENYQVLRFLTAFLVQISAHSDQNKMTNTNLAVVFGPNLLWAKDAAITLKAINPINTFTKFLLDHQGELFPSPDPSGL
Specifications
Specifications
| Antigen | Rho GTPase activating protein 1 |
| Anwendungen | ELISA, Western Blot |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Beschreibung | Mouse polyclonal antibody raised against a partial recombinant ARHGAP1. |
| Zusammensetzung | 50% glycerol |
| Gen | ARHGAP1 |
| Gen-Zugriffsnummer | BC018118 |
| Gen-Alias | CDC42GAP/RHOGAP/RHOGAP1/p50rhoGAP |
| Gensymbole | ARHGAP1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?