missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP13A1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518860
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with canine, mouse and rat based on sequence homology.
ATP13A1 Polyclonal antibody specifically detects ATP13A1 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
ATP13A1 | |
Polyclonal | |
Unconjugated | |
ATP13A1 | |
ATP13A; Atp13a1; ATPase 13A1; ATPase type 13A1; cation transporting ATPase; cation-transporting ATPase 2; catp; Cgi152; CGI-152; DKFZp761L1623; FLJ31858; FLJ41786; FLJ43873; FLJ90317; KIAA1825; Manganese-transporting ATPase 13A1; probable cation-transporting ATPase 13A1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
57130 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9HD20 | |
ATP13A1 | |
Synthetic peptide sequence (REAQARSPEKQEQ) corresponding to the internal amino acids of ATP13A1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |