Learn More
CD273 (B7-DC) Monoclonal Antibody (176611), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA523887
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In direct ELISAs, no cross-reactivity with recombinant mouse PD-L2, recombinant human (rh) B7-1, rhB7-2, rhB7-H1, rhB7-H2, or rhB7-H3 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
CD273 (B7-DC) Monoclonal antibody specifically detects CD273 (B7-DC) in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot.Spezifikation
CD273 (B7-DC) | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9BQ51 | |
PDCD1LG2 | |
Mouse myeloma cell line NS0-derived recombinant human PD-L2/B7-DC Leu20-Pro219 (predicted) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
176611 | |
Unconjugated | |
PDCD1LG2 | |
B7 dendritic cell molecule; B7DC; B7-DC; bA574F11.2; Btdc; Butyrophilin B7-DC; butyrophilin-like protein; CD273; F730015O22Rik; MGC124039; MGC124040; PD1 ligand 2; PD-1 ligand 2; PD-1-ligand 2; PDCD1 ligand 2; PDCD1L2; Pdcd1lg2; Pdl2; PD-L2; Programmed cell death 1 ligand 2; programmed death ligand 2 | |
Mouse | |
Protein A/G | |
RUO | |
80380 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |