missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BACE1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA112529
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
In Western blot, this antibody detects a ∽70 kDa band in human brain lysate.
Accumulation of the amyloid-b (Ab) plaque in the cerebral cortex is a critical event in the pathogenesis Alzheimer′s disease. Ab peptide is generated by proteolytic cleavage of the b-amyloid protein precursor(APP) at b- and g-sites by two proteases. APP is first cleaved by b-secretase, producing a soluble derivative of the protein and a membrane anchored 99-amino acid carboxy-terminal fragment (C99). The C99 fragment serves as substrate for g-secretase to generate the 4 kDa amyloid-b peptide, which is deposited in the brains of all sufferers of Alzheimer′s disease. The long-sought b-secretase was recently identified by several groups independently and designated beta-site APP cleaving enzyme (BACE) and aspartyl protease 2 (Asp2). bACE/Asp2 is a novel transmembrane aspartic protease and colocalizes with APP.Spezifikation
BACE1 | |
Polyclonal | |
Unconjugated | |
BACE1 | |
beta Secretase 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23621, 23821 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P56817, P56818 | |
BACE1 | |
Synthetic peptide corresponding to residues 485-501 of human BACE. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |