Learn More
BACE2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA1753
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-753 detects human beta-secretase 2 (BACE2) from transfected cells and rat and mouse samples. PA1-753 has been successfully used in Western blot and ICC/IF procedures. By Western blot this antibody detects an ∽58 kDa protein representing BACE2 from SY-SY5Y cells overexpressing the human gene. The PA1-753 immunizing peptide corresponds to amino acid residues 441-457 from human BACE2. This sequence is completely conserved in mouse. The PA1-753 immunizing peptide (Cat. # PEP-166) is available for use in neutralization and control experiments.
BACE2 Polyclonal antibody specifically detects BACE2 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
BACE2 | |
Polyclonal | |
Unconjugated | |
BACE2 | |
1110059C24Rik; 56 kDa aspartic-like protease; AEPLC; AI850424; ALP56; ARP1; asp 1; ASP1; ASP21; Aspartic-like protease 56 kDa; Aspartyl protease 1; Bace2; BAE2; be; beta secretase 2; Beta-secretase 2; beta-site amyloid beta A4 precursor protein-cleaving enzyme 2; beta-site amyloid precursor protein cleaving enzyme 2; Beta-site APP cleaving enzyme 2; beta-site APP-cleaving enzyme 2; CDA13; CEAP1; Down region aspartic protease; Down syndrome region aspartic protease; DRAP; memapsin 1; memapsin-1; Membrane-associated aspartic protease 1; theta-secretase; transmembrane aspartic proteinase Asp1; UNQ418/PRO852 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25825, 288227, 56175 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
Q6IE75, Q9JL18, Q9Y5Z0 | |
BACE2 | |
Synthetic Peptide: V(441) S E I S G P F S T E D I A S N C(457) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |