missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bacillus anthracis Lethal Factor Monoclonal Antibody (BAL0106), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA122669
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-22669 detects Bacillus anthracis lethal factor in Bacteria samples.
Bacillus anthracis Lethal Factor Monoclonal antibody specifically detects Bacillus anthracis Lethal Factor in Bacteria samples. It is validated for ELISA.Spezifikation
Bacillus anthracis Lethal Factor | |
Monoclonal | |
2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
Bacteria | |
Antibody | |
IgG1 |
ELISA | |
BAL0106 | |
Unconjugated | |
Anthrax LF; B. anthracis LF | |
Highly purified lethal factor antigen of Bacillus anthracis. | |
100 μg | |
Primary | |
4° C, do not freeze | |
Liquid |