missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAFF (soluble) Monoclonal Antibody (Buffy 2), Invitrogen™
Rat Monoclonal Antibody
Marke: Invitrogen MA191157
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
BAFF (soluble) Monoclonal antibody specifically detects BAFF (soluble) in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
BAFF (soluble) | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q9WU72, Q9Y275 | |
TNFSF13B | |
Recombinant soluble protein, corresponding to amino acids 83 - 285 of Human BAFF. | |
50 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgM |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
Buffy 2 | |
Unconjugated | |
TNFSF13B | |
ApoL related ligand TALL-1; b cell-activating factor; B lymphocyte stimulator; BAFF; B-cell-activating factor; B-lymphocyte stimulator; BLYS; CD257; D8Ertd387e; delta BAFF; Delta4 BAFF; dendritic cell-derived TNF-like molecule; DTL; RGD1561519; TALL1; TALL-1; THANK; TNF- and APOL-related leukocyte expressed ligand 1; TNF and ApoL-related leukocyte expressed ligand 1; TNF homolog that activates apoptosis; Tnfsf13b; TNFSF20; TNLG7A; tumor necrosis factor (ligand) superfamily member 13b; tumor necrosis factor (ligand) superfamily, member 13b; tumor necrosis factor (ligand) superfamily, member 20; tumor necrosis factor ligand 7A; tumor necrosis factor ligand superfamily member 13B; Tumor necrosis factor ligand superfamily member 13b, membrane form; Tumor necrosis factor ligand superfamily member 13b, soluble form; tumor necrosis factor superfamily member 13b; tumor necrosis factor-like protein ZTNF4; UNQ401/PRO738; ZTNF4 | |
Rat | |
Size-exclusion - Gel filtration | |
RUO | |
10673, 24099 | |
4° C | |
Liquid |