missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAG1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA130414
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HL-60.
BAG1 Polyclonal antibody specifically detects BAG1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Spezifikation
BAG1 | |
Polyclonal | |
Unconjugated | |
Bag1 | |
BAG family molecular chaperone regulator 1; Bag1; BAG-1; BCL2 associated athanogene 1; Bcl-2 associating athanogene-1 protein; BCL2-associated athanogene; bcl-2-associated athanogene 1; BCL2-associated athanogene 1; Bcl-2-binding protein; glucocortoid receptor-associated protein RAP46; HAP; Rap46; receptor-associated protein, 46-KD; RP11-326F20.2 | |
Rabbit | |
IgG fraction | |
RUO | |
12017, 297994, 573 | |
4° C, do not freeze | |
Liquid |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 0.2% gelatin and 0.1% sodium azide | |
B0K019, Q60739, Q99933 | |
Bag1 | |
Synthetic peptide (Mouse) (C terminal). | |
1 mL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |