Learn More
BAG3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527696
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: K562, mouse heart. Predicted reactivity: Mouse (94%), Rat (95%), Rhesus Monkey (100%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
BAG3 Polyclonal antibody specifically detects BAG3 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
BAG3 | |
Polyclonal | |
Unconjugated | |
BAG3 | |
AA407278; BAG family molecular chaperone regulator 3; BAG3; BAG-3; BCL2 associated athanogene 3; Bcl-2-associated athanogene 3; Bcl2-associated athanogene 3; BCL2-binding athanogene 3; bcl-2-binding protein Bis; Bcl-2-interacting death suppressor; BIS; CAIR-1; DKFZp434E0610; docking protein CAIR-1; MFM6; mg638; MGC104307; MNCb-2243 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29810, 9531 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.41 mg/mL | |
PBS with 10% glycerol and 0.01% thimerosal; pH 7 | |
O95817, Q9JLV1 | |
BAG3 | |
Recombinant fragment corresponding to a region within amino acids 368 and 567 of Human BAG3 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |