missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAI1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA146465
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Suggested positive control: human or mouse brain.
BAI1 Polyclonal antibody specifically detects BAI1 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
BAI1 | |
Polyclonal | |
Unconjugated | |
ADGRB1 | |
ADGRB1; Adhesion G protein-coupled receptor B1; B830018M07Rik; Bai1; brain-specific angiogenesis inhibitor 1; FLJ41988; GDAIF; mKIAA4089; R75078; Vasculostatin-120; Vasculostatin-40; Vstat120; Vstat40; Vstat-40 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
107831, 575 | |
4° C, do not freeze | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
tris citrate, tris phosphate with 0.1% sodium azide; pH 7-8 | |
O14514, Q3UHD1 | |
ADGRB1 | |
Synthetic peptide corresponding to residues 1500-1584 of C-terminal portion of human BAI1. [Swiss-Prot# O14514] | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |