Learn More
BAI2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA533353
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is not recommended for use in ICC applications. Percent identity with other species by BLAST analysis: Human, Gorilla, Marmoset (100%) Orangutan, Monkey, Horse (95%) Hamster, Elephant, Panda, Pig (89%).
BAI2 Polyclonal antibody specifically detects BAI2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
BAI2 | |
Polyclonal | |
Unconjugated | |
ADGRB2 | |
Adgrb2; Adhesion G protein-coupled receptor B2; BAI2; Brain-specific angiogenesis inhibitor 2; G protein-coupled receptor brain-specific angiogenesis inhibitor 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
576 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
O60241 | |
ADGRB2 | |
Synthetic 19 amino acid peptide from N-terminal extracellular domain of human BAI2. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |