missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGI1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA515496
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with rat based on sequence homology.
MAGI1 Polyclonal antibody specifically detects MAGI1 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
MAGI1 | |
Polyclonal | |
Unconjugated | |
Magi1 | |
AIP3; AIP-3; Atrophin-1-interacting protein 3; BAI1-associated protein 1; Baiap1; BAP1; BAP-1; Gukmi1; MAGI1; Magi-1; MAGI1c; Magi1d; membrane associated guanylate kinase, WW and PDZ domain containing 1; membrane-associated guanylate kinase inverted 1; Membrane-associated guanylate kinase, WW and PDZ domain-containing protein 1; mKIAA4129; TNRC19; Trinucleotide repeat-containing gene 19 protein; WW domain-containing protein 3; WWP3 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
14924, 9223 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
Q6RHR9, Q96QZ7 | |
Magi1 | |
KLH conjugated synthetic peptide between 1227-1256 amino acids from the C-terminal region of human BAIAP1 (MAGI1) | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |