missing translation for 'onlineSavingsMsg'
Learn More

BARHL2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA124446

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11541023

  • 492.00 € / 50 Mikrogramm
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

PA1-24446 detects BARHL2 in Human samples. PA1-24446 has been successfully used in Western Blot procedures. PA1-24446 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human BARHL2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

BARHL2 and BARHL1 are two homeobox genes in mouse and human, which are highly related to the Bar Drosophila genes.
Spezifikation

Spezifikation

BarHL2
Polyclonal
Unconjugated
Q9NY43
Rabbit
Antigen affinity chromatography
RUO
343472
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry, Western Blot
1 mg/mL
BARHL2
BARHL2
Synthetic peptide region of human BARHL2.
50 μg
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote