missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHF11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA541572
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: MERISAFFSS IWDTILTKHQ EGIYNTICLG VLLGLPLLVI ITLLFICCHC Sequence homology: Cow: 92%; Horse: 92%; Human: 100%
PHF11 Polyclonal antibody specifically detects PHF11 in Human samples. It is validated for Western Blot.Spezifikation
PHF11 | |
Polyclonal | |
Unconjugated | |
PHF11 | |
4933417L10Rik; APY; atopic; BCAP; BRCA1 C-terminus-associated protein; BRCA1-C terminus associated protein; D14Ertd668e; IgE responsiveness; IgE responsiveness (atopic); IGEL; IGER; IGHER; MGC137849 protein; NYREN34; NY-REN-34; NY-REN-34 antigen; PHD finger protein 11; PHD finger protein 11 family member; PHD finger protein 11A; PHD finger protein 11D; PHD finger protein 11-like; Phf11; Phf11-4; Phf11a; Phf11d; Phf11l; Renal carcinoma antigen NY-REN-34; RP11-185C18.3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
51131 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UIL8 | |
PHF11 | |
Synthetic peptide directed towards the N-terminal of human PHF11 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |