Learn More
Bcl-2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA520068
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
A suggested positive control is Daudi cell lysate. PA5-20068 can be used with blocking peptide PEP-0187. The PA5-20068 immunogen is located within the first 50 amino acids of Bcl-2. Predicted molecular ∽ 26kD. In Western blot applications, this antibody has been observed to detect a band at: 26 kD. Predicted species reactivity based on immunogen sequence: Bovine: (100%), Rat: (100%), Chicken: (93%)
Bcl-2 Polyclonal antibody specifically detects Bcl-2 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Bcl-2 | |
Polyclonal | |
Unconjugated | |
Bcl2 | |
apoptosis regulator Bcl-2; AW986256; B cell leukemia/lymphoma 2; B cell lymphoma 2 associated oncogene; B-cell CLL/lymphoma 2; B-cell leukemia/lymphoma 2; Bcl 2; BCL2; bcl-2; BCL2 apoptosis regulator; Bcl-2 protein; BCL2, apoptosis regulator; Bcl2-like protein; C430015F12Rik; D630044D05Rik; D830018M01Rik; OTTHUMP00000163680; PPP1R50; protein phosphatase 1, regulatory subunit 50 | |
Rabbit | |
Ion-exchange chromatography | |
RUO | |
12043, 596 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P10415, P10417 | |
Bcl2 | |
A peptide corresponding to 15 amino acids near the N terminus of human Bcl-2. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |