Learn More
Bcl-3 Monoclonal Antibody (Ham150-3.5), Invitrogen™
Armenian Hamster Monoclonal Antibody
Marke: Invitrogen MA1950
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-950 detects human and mouse Bcl-3 from transfected cells and shows no cross-reactivity to other Bcl isoforms. MA1-950 has been successfully used in Western blot and FACS procedures. By Western blot, this antibody detects an ∽47 kDa protein representing Bcl-3 from C57BI/6 human T cells and Bcl-3 transgenic ms T cells. MA1-950 immunogen is full length Bcl-3 protein from mouse.
Bcl-3 Monoclonal antibody specifically detects Bcl-3 in Human, Mouse samples. It is validated for ELISA, Flow Cytometry, Western Blot.Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Spezifikation
Bcl-3 | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
P20749, Q9Z2F6 | |
Bcl3 | |
Full length mouse Bcl-3 protein. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
ELISA, Flow Cytometry, Western Blot | |
Ham150-3.5 | |
Unconjugated | |
Bcl3 | |
AI528691; B cell leukemia/lymphoma 3; B-cell CLL/lymphoma 3; B-cell leukemia/lymphoma 3; B-cell lymphoma 3 protein; B-cell lymphoma 3 protein homolog; B-cell lymphoma 3-encoded protein; BCL3; BCL-3; Bcl-3 protein; BCL4; chronic lymphatic leukemia protein; D19S37; Proto-oncogene BCL3 | |
Armenian Hamster | |
Protein G | |
RUO | |
12051, 602 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |