Learn More
BDH2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA544760
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: NRCVYSTTKA AVIGLTKSVA ADFIQQGIRC NCVCPGTVDT PSLQERIQAR Sequence homology: Cow: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%
BDH2 Polyclonal antibody specifically detects BDH2 in Human samples. It is validated for Western Blot.Spezifikation
BDH2 | |
Polyclonal | |
Unconjugated | |
BDH2 | |
1810026B04Rik; 3-hydroxybutyrate dehydrogenase 2; 3-hydroxybutyrate dehydrogenase type 2; 3-hydroxybutyrate dehydrogenase, type 2; bdh2; dehydrogenase/reductase (SDR family) member 6; dehydrogenase/reductase SDR family member 6; Dhrs6; EFA6R; FLJ13261; hypothetical protein LOC550507; oxidoreductase UCPA; PRO20933; R-beta-hydroxybutyrate dehydrogenase; SDR15C1; Short chain dehydrogenase/reductase family 15C member 1; short chain dehydrogenase/reductase family 15C, member 1; si:dkey-162b23.2; UCPA-OR; UNQ6308; UNQ6308/PRO20933; zgc:110323 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
56898 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BUT1 | |
BDH2 | |
Synthetic peptide directed towards the middle region of human BDH2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |