missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BDNF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA515198
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine, mouse, porcine and rat based on sequence homology.
BDNF Polyclonal antibody specifically detects BDNF in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
BDNF | |
Polyclonal | |
Unconjugated | |
BDNF | |
abrineurin; ANON2; anorexia BDNF; Bdnf; BDNF precursor form; bdnf protein; brain derived neurothrophic factor; brain derived neurotrophic factor; brain-derived neurotrophic factor; BULN2; H-BDNF; neurotrophin; ProBDNF | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
627 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
P23560 | |
BDNF | |
KLH conjugated synthetic peptide between 206-236 amino acids from the C-terminal region of human BDNF | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |