Learn More
beta-2 Microglobulin Monoclonal Antibody (YTH470.5), FITC, Invitrogen™
Rat Monoclonal Antibody
Marke: Invitrogen MA517685
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-17685 targets Beta 2-microglobulin in FACS and WB applications and shows reactivity with Human samples. The MA5-17685 immunogen is rat monoclonal antibody raised against human lymphocytes.
beta-2 Microglobulin Monoclonal antibody specifically detects beta-2 Microglobulin in Human samples. It is validated for Flow Cytometry, Western Blot.Spezifikation
beta-2 Microglobulin | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P61769 | |
B2m | |
Rat monoclonal antibody raised against human lymphocytes. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Western Blot | |
YTH470.5 | |
FITC | |
B2m | |
B2m; B2-microglobulin; b2m-W01; b2m-W03; b2m-Z01; b2m-Z02; b2m-Z03; Beta 2 microglobin; Beta 2 microglobulin; beta 2-microglobulin; beta chain of MHC class I molecules; Beta2 microglobulin; beta-2 microglobulin; beta-2 microglobulin preprotein; beta2m; beta2-m; beta-2-microglobin; beta-2-microglobulin; beta-2-microglobulin C-terminal fragment (55 AA); Beta-2-microglobulin form pI 5.3; B-F; CDABP0092; HDCMA22P; IMD43; Lactollin; Ly-m11; MHC Class I; microglobulin beta-2; Orla-B2m | |
Rat | |
Protein G | |
RUO | |
567 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |