Learn More
CRYBA1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA528954
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, H1299, HeLa, Molt-4, Raji, mouse fetal eye. Predicted reactivity: Mouse (95%), Rat (95%), Xenopus laevis (84%), Dog (96%), Rabbit (96%), Chicken (84%), Rhesus Monkey (97%), Bovine (93%), Guinea pig (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CRYBA1 Polyclonal antibody specifically detects CRYBA1 in Human, Mouse samples. It is validated for Western Blot.Spezifikation
CRYBA1 | |
Polyclonal | |
Unconjugated | |
CRYBA1 | |
BA3/A1; BA3A1C; beta crystallin A3 chain transcript CN; beta crystallin A3 chain transcript LAM; beta crystallin A3 chain transcript PS; beta crystallin A3 chain transcript TC; beta-A1-crystallin; beta-A3; beta-A3 crystallin; beta-A3/A1-crystallin; beta-crystallin A1; beta-crystallin A3; Beta-crystallin A3, isoform A1, Delta4 form; Beta-crystallin A3, isoform A1, Delta7 form; Beta-crystallin A3, isoform A1, Delta8 form; Cryb; CRYB1; Cryba1; crystallin beta A1; crystallin beta A3; crystallin beta A3/A1; crystallin, beta A1; crystallin, beta A3; CTRCT10; eye lens structural protein; truncated beta-crystallin A3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12957, 1411 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.26 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P02525, P05813 | |
CRYBA1 | |
Full length human beta A3 Crystallin Recombinant protein. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |