missing translation for 'onlineSavingsMsg'
Learn More

beta Actin Monoclonal Antibody (15G5A11/E2), Alexa Fluor™ 555, Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MA1140A555

274.68 EUR gültig bis 2024-03-29
Benutzen Sie den Promotions-Code "21615" um Ihren Promotions-Preis zu erhalten.



Weitere Details : Netto-Gewicht : 0.00900kg

Artikelnummer. 15517201

  • 412.00 € / 100 Mikrogramm
Verfügbar ab: 16-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

beta Actin Monoclonal antibody specifically detects beta Actin in Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunocytochemistry, Western Blot.
Spezifikation

Spezifikation

beta Actin
Monoclonal
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
P60709, P60710, P60711
ACTB
Purified full-length recombinant human beta-actin expressed in E. coli
50 μL
Primary
Human, Mouse, Non-human Primate, Rat
Antibody
IgG1
Immunocytochemistry, Western Blot
15G5A11/E2
Alexa Fluor 555
ACTB
0610041G09Rik; AA959943; AAT6; ACT; Act4; Act-4; ACT-5; ACTA; acta1; acta1b; ACTA2; Acta-2; ACTA3; actb; actb.L; actb1; actba; ACTC; actc1; Actc-1; actc1b; ACTE; Actg; ACTG1; Actg2; ACTGE; actin; actin alpha 1; actin alpha 1, skeletal muscle; actin alpha 1, skeletal muscle b; actin alpha 2, smooth muscle; actin alpha cardiac; actin alpha cardiac 1; actin alpha cardiac muscle 1; actin alpha cardiac muscle 1b; actin beta; actin gamma 1; actin gamma 2, smooth muscle; actin, alpha 1, skeletal muscle; actin, alpha 1b, skeletal muscle; actin, alpha 2, smooth muscle, aorta; Actin, alpha cardiac muscle 1; Actin, alpha cardiac muscle 1, intermediate form; actin, alpha cardiac muscle 1b; actin, alpha skeletal muscle; Actin, alpha skeletal muscle, intermediate form; actin, alpha, cardiac 1; actin, alpha, cardiac muscle; actin, alpha, cardiac muscle 1; actin, alpha, vascular smooth muscle; actin, aortic smooth muscle; Actin, aortic smooth muscle, intermediate form; actin, beta; actin, beta 1; acti
Mouse
Protein A
RUO
11461, 60, 81822
4° C, do not freeze
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote