Learn More
beta Actin Monoclonal Antibody (8H10D10), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA515452
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-15452 targets beta-Actin in indirect ELISA, FACS, IF, and WB applications and shows reactivity with Hamster, Human, mouse, Non-human primate, and Rat samples. The MA5-15452 immunogen is synthetic peptide corresponding to amino-terminal residues of human beta-Actin, conjugated to KLH. MA5-15452 detects beta-Actin which has a predicted molecular weight of approximately 42kDa.
beta Actin Monoclonal antibody specifically detects beta Actin in Hamster, Human, Mouse, Non-human Primate, Rat samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Western Blot.Spezifikation
beta Actin | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P48975, P60709, P60710, P60711 | |
ACTB | |
Synthetic peptide corresponding to amino-terminal residues of human beta-Actin, conjugated to KLH. | |
RUO | |
100689477, 101844587, 11461, 60, 81822 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Western Blot | |
8H10D10 | |
Unconjugated | |
ACTB | |
0610041G09Rik; AA959943; AAT6; ACT; Act4; Act-4; ACT-5; ACTA; acta1; acta1b; ACTA2; Acta-2; ACTA3; actb; actb.L; actb1; actba; ACTC; actc1; Actc-1; actc1b; ACTE; Actg; ACTG1; Actg2; ACTGE; actin; actin alpha 1; actin alpha 1, skeletal muscle; actin alpha 1, skeletal muscle b; actin alpha 2, smooth muscle; actin alpha cardiac; actin alpha cardiac 1; actin alpha cardiac muscle 1; actin alpha cardiac muscle 1b; actin beta; actin gamma 1; actin gamma 2, smooth muscle; actin, alpha 1, skeletal muscle; actin, alpha 1b, skeletal muscle; actin, alpha 2, smooth muscle, aorta; Actin, alpha cardiac muscle 1; Actin, alpha cardiac muscle 1, intermediate form; actin, alpha cardiac muscle 1b; actin, alpha skeletal muscle; Actin, alpha skeletal muscle, intermediate form; actin, alpha, cardiac 1; actin, alpha, cardiac muscle; actin, alpha, cardiac muscle 1; actin, alpha, vascular smooth muscle; actin, aortic smooth muscle; Actin, aortic smooth muscle, intermediate form; actin, beta; actin, beta 1; acti | |
Mouse | |
100 μL | |
Primary | |
Hamster, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG2b |