missing translation for 'onlineSavingsMsg'
Learn More

beta-4 Tubulin Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA521416

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 13258968

  • 512.00 € / 100 Mikroliter
Verfügbar ab: 14-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Recommended positive controls: 293T, NIH-3T3. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (96%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

beta-4 Tubulin Polyclonal antibody specifically detects beta-4 Tubulin in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.
Spezifikation

Spezifikation

beta-4 Tubulin
Polyclonal
Unconjugated
TUBB4A
AI325297; beta-5; class IVa beta-tubulin; dystonia 4, torsion (autosomal dominant); DYT4; hypothetical protein LOC540236; M(beta)4; QtrA-11757; Tubb; TUBB4; Tubb4a; TUBB5; Tubulin 5 beta; tubulin beta; tubulin beta 4A class IVa; tubulin beta-4 chain; Tubulin beta-4A chain; tubulin, beta 4; tubulin, beta 4 class IVa; tubulin, beta 4A class IVa; tubulin, beta, 5; unnamed protein product
Rabbit
Antigen affinity chromatography
RUO
10382, 22153, 29213
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Western Blot
0.41 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P04350, Q9D6F9
TUBB4A
Recombinant protein fragment contain a sequence corresponding to a region within amino acids 238 and 410 (P04350) of beta Tubulin 4
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote