Learn More
beta Synuclein Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Marke: Invitrogen PA118260
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Reconstitute in 100 μL of sterile water. Centrifuge to remove any insoluble material. After reconstitution keep aliquots at -20 °C for a higher stability, and at 4 °C with an appropriate antibacterial agent. Glycerol (1:1) may be added for an additional stability. Avoid repetitive freeze/thaw cycles.
beta Synuclein Polyclonal antibody specifically detects beta Synuclein in Human, Rat samples. It is validated for Immunohistochemistry.Spezifikation
beta Synuclein | |
Polyclonal | |
Unconjugated | |
SNCB | |
AI838531; betaSYN; beta-synuclein; Phosphoneuroprotein 14; PNP 14; Sncb; synuclein beta; synuclein, beta | |
Sheep | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Immunohistochemistry | |
Conc. Not Determined | |
whole serum with no preservative | |
Q16143, Q63754 | |
SNCB | |
Synthetic peptide corresponding to residues I(108) E P L M E P E G S Y E D P P Q E(125) of human beta synuclein protein. | |
RUO | |
113893, 6620 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |