missing translation for 'onlineSavingsMsg'
Learn More
Learn More
betacellulin Monoclonal Antibody (34362.11), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA124680
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Does not cross-react with recombinant human EGF, HB-EGF, TGF-alpha HRG-alpha and AR. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
betacellulin Monoclonal antibody specifically detects betacellulin in Human samples. It is validated for ELISA, Western Blot.Spezifikation
betacellulin | |
Monoclonal | |
0.5 mg/mL | |
PBS with no preservative | |
P35070 | |
BTC | |
Recombinant, full length human protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
34362.11 | |
Unconjugated | |
BTC | |
Bcn; betacellulin; betacellulin, epidermal growth factor family member; BTC; OTTHUMP00000160600; OTTHUMP00000219057; Probetacellulin | |
Mouse | |
Protein A | |
RUO | |
685 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |