missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biglycan Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA513700
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine, mouse and rat based on sequence homology.
Biglycan Polyclonal antibody specifically detects Biglycan in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Biglycan | |
Polyclonal | |
Unconjugated | |
BGN | |
BG; BGN; Biglycan; bone/cartilage proteclycan 1; bone/cartilage proteoglycan I; bone/cartilage proteoglycan-I; BSPG1; dermatan sulphate proteoglycan I; DSPG1; PG S1; PGI; PG-S1; SLRR1A; small leucine-rich protein 1A; Small proteoglycan I (biglycan), bone (BSPG1) (bone/cartilage proteclycan 1 precursor) | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
633 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
P21810 | |
BGN | |
KLH conjugated synthetic peptide between 148-176 amino acids from the central region of human BGN | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |