Learn More
BHMT Monoclonal Antibody (8C11H5), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA515465
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-15465 targets BHMT in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15465 immunogen is purified recombinant fragment of BHMT expressed in E. Coli.
BHMT Monoclonal antibody specifically detects BHMT in Human samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Spezifikation
BHMT | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
Q93088 | |
BHMT | |
Purified recombinant fragment of BHMT expressed in E. Coli. | |
RUO | |
635 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
8C11H5 | |
Unconjugated | |
BHMT | |
betaine-homocysteine methyltransferase; betaine--homocysteine S-methyltransferase; betaine-homocysteine S-methyltransferase; betaine--homocysteine S-methyltransferase 1; BHMT; BHMT1; epididymis secretory sperm binding protein Li 61p; HEL-S-61p | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |