missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BIK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA517641
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with other Bcl-2 family members.
BIK Polyclonal antibody specifically detects BIK in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
BIK | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
Q13323 | |
BIK | |
Synthetic peptide corresponding to residues in the amino-terminus of Bik | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
Unconjugated | |
BIK | |
apoptosis inducer NBK; apoptosis-inducing NBK; BBC1; BCL2 interacting killer; bcl-2-interacting killer; BCL2-interacting killer; BCL2-interacting killer (apoptosis-inducing); bhikhari; BIK; Bik-like killer protein; Biklk; BIP 1; BIP1; Blk; BP 4; BP4; cb 60; natural born killer; NBK | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
638 | |
-20°C | |
Liquid |