missing translation for 'onlineSavingsMsg'
Learn More

Bim Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA532276

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 13466797

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen


Beschreibung

Beschreibung

Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is placenta tissue.

BIM/BOD (BCL-2-related Ovarian Death Gene). It belongs to BCL-2 family of proteins containing BCL-2 homology domain3 (BH3). It is proapoptotic and exerts its effects by interacting with prosurvival members of the BCL-2 family like BCL-2, BCL-XL and BCL-W. It exhibits three splice variants; BimL, BimS and BimEL. They all share homology in their C-terminal BH3 domain and are ubiquitously expressed. BimS is the strongest of them with respect to the proapoptotic capacity.
Spezifikation

Spezifikation

Bim
Polyclonal
BCL2L11
BAM, BIM, BIM-alpha6, BIM-beta6, BIM-beta7, BOD, BimEL, BimL, bcl-2 interacting mediator of cell death, bcl-2 interacting protein Bim, bcl-2-like protein 11, bcl-2-related ovarian death agonist, bcl2-L-11
Rabbit
Antigen affinity chromatography
RUO
10018
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
O43521
BCL2L11
Synthetic peptide derived from N-terminus of human BIM.
500 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote