missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bim Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518285
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Bim Polyclonal antibody specifically detects Bim in Human samples. It is validated for Western Blot.Spezifikation
Bim | |
Polyclonal | |
Unconjugated | |
BCL2L11 | |
1500006F24Rik; BAM; Bcl2 interacting mediator of cell death; bcl-2 interacting mediator of cell death; bcl-2 interacting protein Bim; BCL2 like 11; Bcl-2 like 11; Bcl-2 related apoptotic gene product BimL; bcl2-interacting mediator of cell death; BCL2L11; Bcl2-L-11; BCL2-like 11 (apoptosis facilitator); bcl-2-like protein 11; bcl-2-like protein 11-like; bcl-2-related ovarian death agonist; bcl-2-related ovarian death protein; BIM; Bim EL; Bim-A; BIM-alpha6; BIM-beta6; BIM-beta7; BIMEL; BimEL protein; BimL; BOD; OTTHUMP00000197013; OTTHUMP00000204247; Unknown (protein for MGC:134444) | |
Goat | |
Antigen affinity chromatography | |
RUO | |
10018 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O43521 | |
BCL2L11 | |
Synthetic peptide sequence (FNAYYARRLEK) corresponding to the C-terminus amino acids of BCL2L11 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |