Learn More
BIN1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA534687
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A431, mouse kidney. Predicted reactivity: Mouse (92%), Rat (80%), Pig (92%), Chicken (81%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
BIN1 Polyclonal antibody specifically detects BIN1 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
BIN1 | |
Polyclonal | |
Unconjugated | |
BIN1 | |
ALP-1; AMPH 2; Amph2; amphiphysin 2; amphiphysin II; amphiphysin IIamph2; amphiphysin-like; amphiphysin-like protein; AMPHL; Bin1; box dependant MYC interacting protein 1; box-dependent myc-interacting protein 1; BRAMP-2; bridging integrator 1; DKFZp547F068; MGC10367; MGC105358; myc box dependent interacting protein 1; myc box-dependent-interacting protein 1; SH3 domain-containing protein 9; SH3P9; similar to bridging integrator 1 isoform 9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
274, 30948 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
2.35 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
O00499, O08539 | |
BIN1 | |
Recombinant protein fragment contains a sequence corresponding to a region within amino acids 1 and 290 of BIN1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |