missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BPNT1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518787
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with mouse and rat based on sequence homology.
BPNT1 Polyclonal antibody specifically detects BPNT1 in Human samples. It is validated for Western Blot.Spezifikation
BPNT1 | |
Polyclonal | |
Unconjugated | |
Bpnt1 | |
3'(2'), 5'-bisphosphate nucleotidase 1; 3'(2'),5'-bisphosphate nucleotidase 1; Bisphosphate 3'-nucleotidase 1; BPNT1; BPntase; epididymis luminal protein 20; HEL20; PAP-inositol 1,4-phosphatase; PAP-inositol-1,4-phosphatase; PIP; Sal3; scHAL2 analogous 3; testicular tissue protein Li 29 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
10380 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O95861 | |
Bpnt1 | |
Synthetic peptide sequence (RNYDYYASRVPESIK) corresponding to the C-terminus amino acids of BPNT1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |