missing translation for 'onlineSavingsMsg'
Learn More

KCNMB2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA539067

Weitere Details : Netto-Gewicht : 0.00900kg

Artikelnummer. 15515981

  • 472.00 € / 100 Mikrogramm
Verfügbar ab: 23-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

MaxiK channels are large conductance, voltage and calcium-sensitive potassium channels which are fundamental to the control of smooth muscle tone and neuronal excitability. MaxiK channels can be formed by 2 subunits: the pore-forming alpha subunit and the modulatory beta subunit. The protein encoded by this gene is an auxiliary beta subunit which decreases the activation time of MaxiK alpha subunit currents. Alternative splicing results in multiple transcript variants of this gene. Additional variants are discussed in the literature, but their full length nature has not been described. [provided by RefSeq, Jul 2013].
Spezifikation

Spezifikation

KCNMB2
Polyclonal
Unconjugated
KCNMB2
KCNMB2
A synthetic peptide derived from the internal region of human KCNMB2
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Western Blot
1 mg/mL
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Q9Y691, Q9CZM9, Q811Q0
Rabbit
Antigen affinity chromatography
RUO
10242, 294961, 72413
-20°C
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote