missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Blood Group A Antigen Monoclonal Antibody (HE-195), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA135443
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Blood Group A Antigen Monoclonal antibody specifically detects Blood Group A Antigen in Human samples. It is validated for Agglutination.Spezifikation
Blood Group A Antigen | |
Monoclonal | |
Conc. Not Determined | |
tissue culture supernatant with <0.1% sodium azide | |
P16442 | |
ABO | |
Mixture of erythrocytes of group A1 and glycoprotein fraction isolated from saliva of secretors with blood group A. | |
RUO | |
28 | |
4° C | |
Liquid |
Agglutination | |
HE-195 | |
Unconjugated | |
ABO | |
A transferase; A1-specific alpha 1-3-N-acetylgalactosaminyltransferase; A3GALNT; A3GALT1; ABO; ABO A3 transferase; ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase; transferase B, alpha 1-3-galactosyltransferase); ABO glycosyltransferase; ABO weak transfer; B transferase; B(A) alpha-1,3-galactosyltransferase; Blood Group A1; Blood Group A2; Blood Group A3; fucosylglycoprotein; fucosylglycoprotein 3-alpha-galactosyltransferase; Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase; Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase soluble form; Glycoprotein-fucosylgalactoside alpha-galactosyltransferase; glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase; glycosyltransferase A; glycosyltransferase B; GTB; histo-blood group A transferase; histo-blood group A2 transferase; histo-blood group ABO system transferase; Histo-blood group B transferase; NAGAT; Transferase; transferase A | |
Mouse | |
1 mL | |
Primary | |
Human | |
Antibody | |
IgM |