missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Blood Group A1B Antigen Monoclonal Antibody (HE-24), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA121745
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-21745 detects Blood Group A1B Antigen in Human samples.
Blood Group A1B Antigen Monoclonal antibody specifically detects Blood Group A1B Antigen in Human samples. It is validated for Agglutination.Spezifikation
Blood Group A1B Antigen | |
Monoclonal | |
Conc. Not Determined | |
tissue culture supernatant with no preservative | |
Mouse | |
1 mL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Agglutination | |
HE-24 | |
Unconjugated | |
Blood Group A | |
Glycoprotein fraction isolated from the saliva of secretors with blood group A. | |
RUO | |
Human | |
Antibody | |
IgM |