Learn More
BLZF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA541466
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: DPVTCKESSP DNPFFESSPT TLLATKKNIG RFHPYTRYEN ITFNCCNHCR Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 93%
BLZF1 Polyclonal antibody specifically detects BLZF1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
BLZF1 | |
Polyclonal | |
Unconjugated | |
BLZF1 | |
1700030G05Rik; Basic leucine zipper nuclear factor 1; basic leucine zipper nuclear factor 1 (JEM-1); Blzf1; blzf1 protein; Blzf1l; cytoplasmic protein; fe06a04; Golgin45; Golgin-45; Golgin-45; golgin-45; JEM1; JEM-1; JEM-1s; JEM-1short protein; LOC504820 protein; MGC22497; p45 basic leucine-zipper nuclear factor; wu:fe06a04; zgc:66280; zgc:85871 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
8548 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H2G9 | |
BLZF1 | |
Synthetic peptide directed towards the C-terminal of human BLZF1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |