Learn More
BMI-1, clone: 1H6B10G7, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen 375400
Weitere Details : Netto-Gewicht : 0.50000kg
Beschreibung
Component of the Polycomb group (PcG) multiprotein PRC1 complex, a complex required to maintain the transcriptionally repressive state of many genes, including Hox genes, throughout development. PcG PRC1 complex acts via chromatin remodeling and modification of histones; it mediates monoubiquitination of histone H2A 'Lys-119', rendering chromatin heritably changed in its expressibility. In the PRC1 complex, it is required to stimulate the E3 ubiquitin-protein ligase activity of RNF2/RING2.
Spezifikation
Bmi-1 | |
Monoclonal | |
0.5mg/mL | |
BMI1 | |
BMI1, PCGF4, RNF51, Polycomb complex protein BMI-1, Polycomb group RING finger protein 4, RING finger protein 51 | |
Mouse | |
Protein A | |
RUO | |
648 | |
-20°C | |
Liquid |
ELISA, Western Blot | |
1H6B10G7 | |
Unconjugated | |
P35226 | |
BMI1 | |
Synthetic peptide derived from the mid region of the human Bim-1 protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |