missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMI-1 Polyclonal Antibody, DyLight™ 550, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522848
Additional Details : Netto-Gewicht : 0.01000kg
Description
Suggested positive control: human and mouse skeletal muscle.
BMI-1 Polyclonal antibody specifically detects BMI-1 in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Specifications
BMI-1 | |
Polyclonal | |
DyLight 550 | |
BMI1 | |
AW546694; B lymphoma Mo-MLV insertion region 1; B lymphoma Mo-MLV insertion region 1 homolog; Bmi1; Bmi-1; BMI1 polycomb ring finger oncogene; BMI1 polycomb ring finger proto-oncogene; BMI1 proto-oncogene, polycomb ring finger; FLVI2/BMI1; flvi-2/bmi-1; murine leukemia viral (bmi-1) oncogene homolog; Pcgf4; Pcgf4 antibody; Polycomb complex protein BMI-1; polycomb group protein Bmi1; polycomb group ring finger 4; polycomb group RING finger protein 4; RING finger protein 51; RNF51; RP11-573G6.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12151, 648 | |
4° C, store in dark | |
Liquid |
Immunocytochemistry, Western Blot | |
0.83 mg/mL | |
50mM sodium borate with no preservative | |
P25916, P35226 | |
BMI1 | |
A synthetic peptide made to an internal region (within residues 1-100) of the human Bmi1 protein. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |