Learn More
BMP-4 Monoclonal Antibody (10F4B4), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA515572
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-15572 targets BMP4 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15572 immunogen is purified recombinant fragment of human BMP4 expressed in E. Coli. MA5-15572 detects BMP4 which has a predicted molecular weight of approximately 64kDa.
BMP-4 Monoclonal antibody specifically detects BMP-4 in Human samples. It is validated for ELISA, Western Blot.Spezifikation
BMP-4 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P12644 | |
BMP4 | |
Purified recombinant fragment of human BMP4 expressed in E. Coli. | |
RUO | |
652 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
10F4B4 | |
Unconjugated | |
BMP4 | |
BMP; BMP2B; BMP-2B; Bmp2b1; Bmp2b-1; Bmp4; Bmp-4; BOMPR4A; Bone morphogenetic protein; Bone morphogenetic protein 2B; bone morphogenetic protein 4; bone morphogenetic protein 4 preproprotein; DVR4; Dvr-4; MCOPS6; OFC11; ZYME | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |