Learn More
BNIP3 Rabbit anti-Human, Biotin, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA522859
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
In Western blot a band is seen in hypoxic samples at ∽30 and 60 kDa, representing the BNIP3 protein. There are some non-specific bands present as well. It is recommended that normoxic samples are run next to the hypoxic samples as a negative control.
This gene is a member of the BCL2/adenovirus E1B 19 kd-interacting protein (BNIP) family. It interacts with the E1B 19 kDa protein which is responsible for the protection of virally-induced cell death, as well as E1B 19 kDa-like sequences of BCL2, also an apoptotic protector. This gene contains a BH3 domain and a transmembrane domain, which have been associated with pro-apoptotic function. The dimeric mitochondrial protein encoded by this gene is known to induce apoptosis, even in the presence of BCL2.Spezifikation
BNIP3 | |
Polyclonal | |
Biotin | |
Q12983 | |
BNIP3 | |
A synthetic peptide against an N-terminal region (within residues 1-50) of the human BNIP3 protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Western Blot | |
1.0 mg/mL | |
BNIP3 | |
NIP3, BCL2/adenovirus E1B 19 kDa protein-interacting protein 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
664 | |
4° C, store in dark | |
Liquid |