Learn More
BNP Monoclonal Antibody (9H6B4,9H6H4A7), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA515362
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-15362 targets BNP in indirect ELISA, IHC applications and shows reactivity with Human samples. The MA5-15362 immunogen is synthetic peptide corresponding to aa (Gly-Leu-Gln-Glu-Gln-Arg-Asn-His-Leu-Gln-Gly-Lys-Leu-Cys) of human BNP, conjugated to KLH.
BNP Monoclonal antibody specifically detects BNP in Human samples. It is validated for ELISA, Immunohistochemistry.Spezifikation
BNP | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P16860 | |
Nppb | |
Synthetic peptide corresponding to aa (Gly-Leu-Gln-Glu-Gln-Arg-Asn-His-Leu-Gln-Gly-Lys-Leu-Cys) of human BNP, conjugated to KLH. | |
RUO | |
4879 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry | |
9H6B4,9H6H4A7, 9H6H4A7 | |
Unconjugated | |
Nppb | |
5 kDa cardiac natriuretic peptide; AA408272; Bnf; BNP; BNP(1-28); BNP(1-29); BNP(1-30); BNP(1-32); BNP(2-31); BNP(3-29); BNP(3-30); BNP(3-32); BNP(4-27); BNP(4-29); BNP(4-30); BNP(4-31); BNP(4-32); BNP(5-29); BNP(5-31); BNP(5-32); BNP-32; BNP-34; Brain natriuretic factor; brain natriuretic peptide; Brain natriuretic peptide 32; Brain natriuretic peptide 45; brain type natriuretic peptide; gamma-brain natriuretic peptide; Iso-ANP; natriuretic peptide B; natriuretic peptide precursor B; natriuretic peptide precursor type B; natriuretic peptide type B; natriuretic peptides B; natriuretic protein; Nppb; type-B natriuretic factor | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |