Learn More
Borrelia burgdorferi garinii Monoclonal Antibody (Bg14), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA121681
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-21681 detects Borrelia burgdorferi garinii in Bacteria samples. MA1-21681 has been successfully used in ELISA, Immunofluorescence and Western Blot procedures. MA1-21681 immunogen corresponds to Bacterial lysates of Borrelia burgdorferi garinii..
Borrelia burgdorferi is a spirochete and the cause of Lyme disease, a tick transmitted illness of humans and animals. B. burgdorferi may persist in humans and animals for months or years following initial infection, despite a robust humoral immune response. B. burgdorferi resembles other spirochetes in that it is a highly specialized, motile, two-membrane, spiral shaped bacteria which lives primarily as an extracellular pathogen. B. burgdorferi has an unusual genome compared with other eubacteria which includes a linear chromosome approximately one megabase in size and numerous linear and circular plasmids.Spezifikation
Borrelia burgdorferi garinii | |
Monoclonal | |
2.8 mg/mL | |
B. burgdorferi; Lyme disease; Borrelia | |
Bacterial lysates of Borrelia burgdorferi garinii. | |
500 μg | |
Primary | |
4° C, do not freeze | |
Liquid |
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot | |
Bg14 | |
Unconjugated | |
Mouse | |
Protein G | |
RUO | |
Bacteria | |
Antibody | |
IgG1 |