Learn More
Borrelia burgdorferi Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA173004
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The PA1-73004 antibody reacts with Borrelia burgdorferi. This antibody cross-reacts with Treponema pallidum, B. hermsii & B. parkerii. PA1-73004 has been successfully used in Western blot, Immunohistochemistry (paraffin), and immunofluorescence applications. By Western blot, this antibody detects bands at 83, 41, 34 & 31 kDa, plus additional low MW bands. For immunofluorescence, specimens should be acetone-fixed for best results. The PA1-73004 antibody was raised against B. burgdorferi whole cell preparation. Short term, less than 6 months, storage is liquid at 4oC. For longer term storage, aliquot and freeze.
Borrelia burgdorferi Polyclonal antibody specifically detects Borrelia burgdorferi in Bacteria samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Borrelia burgdorferi | |
Polyclonal | |
Unconjugated | |
B. burgdorferi; Borrelia; Lyme disease | |
B. burgdorferi whole cell preparation. | |
1 mL | |
Primary | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
4-5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Rabbit | |
purified | |
RUO | |
Bacteria | |
Antibody | |
IgG |