missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bovine Papilloma Virus Type 1 E2 Monoclonal Antibody (1E2), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA171534
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody detects BPV type 1 E2 protein, amino acids 182-190 (TTGHYSVRD).
Bovine Papilloma Virus Type 1 E2 Monoclonal antibody specifically detects Bovine Papilloma Virus Type 1 E2 in Virus samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot.Spezifikation
Bovine Papilloma Virus Type 1 E2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and no preservative; pH 7.4 | |
P03122 | |
E2 | |
Recombinant full length BPV type I E2 protein purified from E. coli | |
100 μg | |
Primary | |
Virus | |
Antibody | |
IgG |
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
1E2 | |
Unconjugated | |
E2 | |
BPV; BPV Protein E2; BPV-1; BPV1-E2 | |
Mouse | |
Protein A | |
RUO | |
1489020 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |